
2,5-Dimethoxyaniline ≥98.0% (by GC, titration analysis)
: TCI
:102-56-7
:
:C₈H₁₁NO₂
:MFCD00008368
:153.18 g/mol
:159°C
:81 °C
:Ambient
:203-040-9
:2811
:6.1,III
:GHS06
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:80.0 to 83.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together