Order Entry
Italy
ContactUsLinkComponent
2,5-Diethoxyaniline ≥98.0% (by GC, titration analysis)
Irritant
2,5-Diethoxyaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0450-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,5-Diethoxyaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0450-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    83.0 to 87.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :181.23 g/mol
Boiling Pt:190 - 192 °C (30 -rr)
Melting Pt:86 °C
EINECS:202-369-5
ADR:6.1,III
 :GHS07
CAS Number:94-85-9
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD00015144