
2,4-Dimethoxyaniline ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2735-04-8
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00008371
:153.18 g/mol
Boiling Pt:262 °C (1013 hPa)
Melting Pt:39 °C
Flash Pt:≥ 130 °C
Storage Temperature:Ambient
EINECS:220-355-7
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to lump
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together