Order Entry
Italy
ContactUsLinkComponent
2,6-Dinitroaniline ≥95.0% (by GC)
Toxic
Health
Environment
2,6-Dinitroaniline ≥95.0% (by GC)
Catalog # TCIAD0815-10G
Supplier:  TCI
CAS Number:  
undefined
Toxic
Health
Environment
2,6-Dinitroaniline ≥95.0% (by GC)
Catalog # TCIAD0815-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    136.0 to 141.0 deg-C

Specification Test Results

About this item

 :
 :183.12 g/mol
Merck Index:13,03299
Melting Pt:136 °C
Density:1,629 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-108-1
UN:1596
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:606-22-4
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007148