Order Entry
Italy
ContactUsLinkComponent
Methyl orange ≥98.0% (by HPLC)
Toxic
Methyl orange ≥98.0% (by HPLC)
Catalog # TCIAM0489-25G
Supplier:  TCI
CAS Number:  
undefined
Toxic
Methyl orange ≥98.0% (by HPLC)
Catalog # TCIAM0489-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Lambda max.:
    460.0 to 470.0 nm (pH 4.8 buffer sol.)
  • Absorbance:
    min. 0.330 (5mg/L, pH 4.8 buffer sol., 460.0 to 470.0 nm) (after drying)
  • Drying loss:
    max. 10.0 %

Specification Test Results

About this item

 :
 :327.34 g/mol
Merck Index:14,06105
Boiling Pt:~ 90 °C (1013 hPa)
Melting Pt:> 300 °C
Density:~ 1 g/cm³ (20 °C)
Flash Pt:38 °C
Storage Temperature:Ambient
EINECS:208-925-3
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502