


2,4-Difluorophenol ≥98.0% (by GC)
: TCI
:367-27-1
:
:C₆H₄F₂O
:MFCD00009715
:130.09 g/mol
:52°C
:20 °C
:1,36 g/cm³ (20 °C)
:206-688-0
:1325
:4.1,III
:GHS02
:GHS05
:GHS07
- Appearance:Light yellow to Yellow powder to lump
- Purity (GC):min. 98.0 %
- Freezing point:21.0 to 24.0 deg-C
Frequently Bought Together