Order Entry
Italy
ContactUsLinkComponent
 
 
 
2,3-Difluorophenol ≥98.0% (by GC)
 :  TCI
 
undefined
 
 
2,3-Difluorophenol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    37.0 to 41.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together