

2,6-Difluoroanisole ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:437-82-1
:
Formula:C₇H₆F₂O
MDL Number:MFCD00142846
:144.12 g/mol
Boiling Pt:50°C
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together