Order Entry
Italy
ContactUsLinkComponent
 
 
2,3-Difluoroanisole ≥97.0% (by GC)
  TCIAD3359-1G
 :  TCI
 :  
undefined
 
2,3-Difluoroanisole ≥97.0% (by GC)
  TCIAD3359-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :144.12 g/mol
 :1993
 :3,III
 :GHS02
 :134364-69-5
 :C₇H₆F₂O
 :MFCD01631562