
3,4-Difluoroaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3863-11-4
:
Formula:C₆H₅F₂N
MDL Number:MFCD00007761
:129.11 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:22 °C
Density:1,302 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:223-381-7
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together