
2,4-Difluoroaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:367-25-9
:
Formula:C₆H₅F₂N
MDL Number:MFCD00007648
:129.11 g/mol
Merck Index:14,03147
Boiling Pt:170°C
Melting Pt:–7 °C
Density:1,268 g/cm³ (20 °C)
Flash Pt:62°C
Storage Temperature:Ambient
EINECS:206-687-5
UN:2941
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together