
2-Fluoro-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:403-24-7
:
Formula:C₇H₄FNO₄
MDL Number:MFCD00275565
:185.11 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:172.0 to 176.0 deg-C
Specification Test Results
Frequently Bought Together