
2,6-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1629-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:157.0 to 161.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:158.1 g/mol
Boiling Pt:72 - 77 °C (13 -rr)
Melting Pt:159…160 °C
EINECS:206-856-3
:GHS07
Storage Temperature:Ambient
CAS Number:385-00-2
Formula:C₇H₄F₂O₂
MDL Number:MFCD00002411