
2,6-Difluorobenzonitrile ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1897-52-5
:
Formula:C₇H₃F₂N
MDL Number:MFCD00001778
:139.1 g/mol
Boiling Pt:98 - 100 °C (31 -rr)
Melting Pt:30…32 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:217-589-7
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together