
1,3-Difluorobenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:372-18-9
:
Formula:C₆H₄F₂
MDL Number:MFCD00000327
:114.09 g/mol
Boiling Pt:83°C
Melting Pt:–59°C
Density:1,16 g/cm³ (20 °C)
Flash Pt:–5°C
Storage Temperature:Ambient
EINECS:206-746-5
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together