Order Entry
Italy
ContactUsLinkComponent
 
 
2,3-Dichlorothiophenol ≥95.0% (by GC)
 :  TCI
 
undefined
 
2,3-Dichlorothiophenol ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    56.0 to 60.0 deg-C

 

Frequently Bought Together