

2,3-Dichlorophenol ≥99.0% (by GC)
Catalog # TCIAD0391-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:58.0 to 61.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:163 g/mol
Boiling Pt:90°C
Melting Pt:57…58 °C
Density:1,4 g/cm³ (25 °C)
Flash Pt:115 °C
Storage Temperature:Ambient
EINECS:209-399-8
UN:2020
ADR:6.1,III
:GHS07
:GHS09
CAS Number:576-24-9
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002160