
4-Chloro-2-nitrophenol ≥98.0% (by GC, titration analysis)
: TCI
:89-64-5
:
:C₆H₄ClNO₃
:MFCD00007113
:173.56 g/mol
:85…88 °C
:Ambient
:201-927-5
:6.1,III
:GHS07
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:86.0 to 89.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together