
5-Nitro-1,2,4-trichlorobenzene ≥99.0% (by GC)
TCIAT0387-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:57.0 to 59.0 deg-C
- Solubility in Methanol:almost transparency
:
:226.45 g/mol
:150°C
:59 °C
:1,79 g/cm³ (22 °C)
:201-931-7
:6.1,III
:GHS07
:89-69-0
:C₆H₂Cl₃NO₂
:MFCD00007072