Order Entry
Italy
ContactUsLinkComponent
1,4-Dichloro-2-iodobenzene ≥98.0% (by GC)
Irritant
1,4-Dichloro-2-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD1435-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,4-Dichloro-2-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD1435-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:255 °C (1013 hPa)
Melting Pt:20 °C
Density:2,02 g/cm³ (20 °C)
Flash Pt:94°C
 :272.9 g/mol
EINECS:249-774-3
 :GHS07
CAS Number:29682-41-5
Formula:C₆H₃Cl₂I
MDL Number:MFCD00001036