Order Entry
Italy
ContactUsLinkComponent
1,3-Dichloro-4-iodobenzene ≥98.0% (by GC)
Irritant
1,3-Dichloro-4-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD1441-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,3-Dichloro-4-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD1441-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :272.9 g/mol
Boiling Pt:262 °C (1013 hPa)
Density:2,01 g/cm³ (20 °C)
EINECS:249-938-4
 :GHS07
CAS Number:29898-32-6
Formula:C₆H₃Cl₂I
MDL Number:MFCD00001035