
4-Chloro-2-fluorobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:446-30-0
:
Formula:C₇H₄ClFO₂
MDL Number:MFCD00042468
:174.56 g/mol
EINECS:207-162-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:205.0 to 209.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together