
2-Chloro-4-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2252-51-9
:
Formula:C₇H₄ClFO₂
MDL Number:MFCD00010615
:174.56 g/mol
EINECS:218-845-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:184.0 to 187.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together