Order Entry
Italy
ContactUsLinkComponent
 
 
 
4-Chloro-1,3-difluorobenzene ≥98.0% (by GC)
  TCIAC1830-25G
 :  TCI
 :  
undefined
 
 
4-Chloro-1,3-difluorobenzene ≥98.0% (by GC)
  TCIAC1830-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :148.54 g/mol
 :127°C
 :-26°C
 :–26°C
 :1,353 g/cm³ (20 °C)
 :32°C
 :1993
 :3,III
 :GHS02
 :GHS07
 :1435-44-5
 :C₆H₃ClF₂
 :MFCD00042569