
3-Amino-4-chlorobenzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2840-28-0
:
Formula:C₇H₆ClNO₂
MDL Number:MFCD00007671
:171.58 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:214…215 °C
Storage Temperature:Ambient
EINECS:220-635-9
:GHS07
Specification Test Results
- Appearance:White to Green to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:213.0 to 216.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together