
3-Chlorobenzoic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:535-80-8
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002491
:156.57 g/mol
Merck Index:14,02123
Boiling Pt:281 °C (1013 hPa)
Melting Pt:122 °C
Density:1,496 g/cm³ (20 °C)
EINECS:208-618-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:155.0 to 159.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together