
2-Chlorobenzaldehyde ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:89-98-5
:
Formula:C₇H₅ClO
MDL Number:MFCD00003304
:140.57 g/mol
Boiling Pt:212°C
Melting Pt:10 °C
Density:1,248 g/cm³ (20 °C)
Flash Pt:104°C
Storage Temperature:Ambient
EINECS:201-956-3
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together