
2,6-Dichlorobenzaldehyde ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:83-38-5
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003307
:175.01 g/mol
Boiling Pt:231°C
Melting Pt:69…70 °C
Flash Pt:≥ 100 °C
Storage Temperature:Ambient
EINECS:201-472-2
UN:1759
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:68.0 to 72.0 deg-C
Specification Test Results
Frequently Bought Together