

2,4-Dichlorobenzaldehyde ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:874-42-0
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003305
:175.01 g/mol
Boiling Pt:233 °C (1013 hPa)
Melting Pt:68…71 °C
Density:1,33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-861-1
UN:1759
ADR:8,III
:GHS05
:GHS09
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:71.0 to 74.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together