Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
Chlorobenzene ≥98.0% (by GC)
  TCIAC1948-500G
 :  TCI
 :  
undefined
 
 
 
Chlorobenzene ≥98.0% (by GC)
  TCIAC1948-500G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :112.56 g/mol
 :14,02121
 :132°C
 :-45°C
 :–45°C
 :1,11 g/cm³ (20 °C)
 :28°C
 :Ambient
 :203-628-5
 :1134
 :3,III
 :01-2119432722-45
 :GHS02
 :GHS07
 :GHS09
 :108-90-7
 :C₆H₅Cl
 :MFCD00000530