
3,5-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1132-21-4
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002502
:182.18 g/mol
Boiling Pt:341 °C (1013 hPa)
Melting Pt:179…182 °C
Density:1,423 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-473-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:182.0 to 187.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together