
2,4-Dimethoxybenzoic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:91-52-1
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002434
:182.18 g/mol
Boiling Pt:313 °C (1013 hPa)
Melting Pt:107…110 °C
Storage Temperature:Ambiente
EINECS:202-074-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:108.0 to 112.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together