
4-Isopropoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13205-46-4
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00044318
:180.2 g/mol
Boiling Pt:305 °C (1013 hPa)
Melting Pt:163…166 °C
Storage Temperature:Ambient
EINECS:236-169-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:163.0 to 167.0 deg-C
Specification Test Results
Frequently Bought Together