
4,5-Dimethoxyanthranilic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD2013-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:197.19 g/mol
Boiling Pt:359 °C (1013 hPa)
Melting Pt:155…160 °C
EINECS:227-095-3
:GHS07
Storage Temperature:Ambient
CAS Number:5653-40-7
Formula:C₉H₁₁NO₄
MDL Number:MFCD00011671