
Syringaldehyde ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:134-96-3
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00006943
:182.18 g/mol
Merck Index:12,09191
Boiling Pt:322 °C (1013 hPa)
Melting Pt:107 °C
Storage Temperature:Ambient
EINECS:205-167-5
:GHS07
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:111.0 to 113.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together