
2,3-Dimethoxybenzaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:86-51-1
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00003309
:166.18 g/mol
Boiling Pt:256°C
Melting Pt:48…49 °C
Storage Temperature:Ambient
EINECS:201-677-7
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 55.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together