
3-Hydroxy-p-anisaldehyde ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:621-59-0
:
Formula:C₈H₈O₃
MDL Number:MFCD00003369
:152.15 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1,401 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-694-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Oxime Formation):min. 98.0 %
- Melting point:112.0 to 116.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together