
2-Bromo-1,4-dimethoxybenzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:25245-34-5
:
Formula:C₈H₉BrO₂
MDL Number:MFCD00008355
:217.06 g/mol
Boiling Pt:237°C
Density:1,445 g/cm³ (15 °C)
EINECS:246-756-7
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together