
2,6-Dibromo-4-fluorophenol ≥98.0% (by GC, titration analysis)
: TCI
:344-20-7
:
:C₆H₃Br₂FO
:MFCD00002317
:269.9 g/mol
:206-451-1
:GHS07
:Ambient
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:54.0 and le 58.0 deg-C
Frequently Bought Together