
1,3-Dibromo-5-fluoro-2-iodobenzene ≥98.0% (by GC)
Codice catalogo TCIAD3582-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:133.0 to 138.0 deg-C
Specification Test Results
About this item
:
Boiling Pt:286 °C (1013 hPa)
Melting Pt:135…135,6 °C
:379.79 g/mol
:GHS07
CAS Number:62720-29-0
Formula:C₆H₂Br₂FI
MDL Number:MFCD00042232