
2-Bromo-4-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1006-41-3
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD00055370
:219.01 g/mol
Boiling Pt:289 °C (1013 hPa)
Melting Pt:173…175 °C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:172.0 to 176.0 deg-C
Specification Test Results
Frequently Bought Together