
2,4-Dichloro-5-methoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:98446-49-2
:
Formula:C₇H₇Cl₂NO
MDL Number:MFCD00974410
:192.04 g/mol
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:49.0 to 53.0 deg-C
Specification Test Results
Frequently Bought Together