
2-Fluorophenylhydrazinium chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2924-15-4
:
Formula:C₆H₈ClFN₂
MDL Number:MFCD00012927
:162.59 g/mol
EINECS:220-885-9
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Potassium iodate Method):min. 97.0 %
Specification Test Results
Frequently Bought Together