

2,6-Toluenediisocyanate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:91-08-7
:
Formula:C₉H₆N₂O₂
MDL Number:MFCD00002010
:174.16
Boiling Pt:103°C
Flash Pt:127°C
Storage Temperature:Refrigerator
EINECS:202-039-0
UN:2078
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together