

1-Bromo-2,4-difluorobenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:348-57-2
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00000330
:192.99 g/mol
Boiling Pt:146°C
Melting Pt:–4 °C
Density:1,69 g/cm³ (20 °C)
Flash Pt:51°C
Storage Temperature:Refrigerator
EINECS:206-479-4
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together