Order Entry
Italy
ContactUsLinkComponent
 
 
 
1,2-Dibromo-4,5-difluorobenzene ≥97.0% (by GC)
 :  TCI
 
undefined
 
 
1,2-Dibromo-4,5-difluorobenzene ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together