
3,5-Dibromosalicylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3147-55-5
:
Formula:C₇H₄Br₂O₃
MDL Number:MFCD00002441
:295.91 g/mol
Merck Index:14,03028
Boiling Pt:365 °C (1013 hPa)
Melting Pt:228…229 °C
Storage Temperature:Ambient
EINECS:221-570-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:222.0 to 230.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together