
3,5-Dibromobenzoic acid ≥97.0% (by GC, titration analysis)
: TCI
:618-58-6
:
:C₇H₄Br₂O₂
:MFCD00051758
:279.92 g/mol
:GHS07
:Ambient
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Solubility in Methanol:very faint turbidity
Frequently Bought Together