
2-Bromobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:88-65-3
:
Formula:C₇H₅BrO₂
MDL Number:MFCD00002402
:201.02 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:144…147 °C
Density:1,929 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
EINECS:201-848-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:146.0 to 152.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together