
2,5-Dibromobenzoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:610-71-9
:
Formula:C₇H₄Br₂O₂
MDL Number:MFCD00016494
:279.92 g/mol
EINECS:210-234-7
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:155.0 to 159.0 deg-C
Specification Test Results
Frequently Bought Together